Anti-Myosin light chain 3 Rabbit Monoclonal Antibody [clone: EPR4160]
Fournisseur: Abcam
New Product
Certificats
À propos de cet article
Rabbit monoclonal [EPR4160] to Myosin light chain 3.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Myosin light chain 3
Clonality: Monoclonal
Clone: EPR4160
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- ABCA_AB108923-100U
- ABCA_AB108923-1ML
- Antigen symbol:
- Myosin light chain 3
- Myosin light chain 3
- Antigen name:
- Cardiac myosin light chain 1
- Cardiac myosin light chain 1
- Antigen synonyms:
- Ventricular myosin alkali light chain|Ventricular myosin light chain 1|Ventricular/slow twitch myosin alkali light chain|CMLC1|MLC1SB|VLCl|MLC-lV/sb|MYL3
- Ventricular myosin alkali light chain|Ventricular myosin light chain 1|Ventricular/slow twitch myosin alkali light chain|CMLC1|MLC1SB|VLCl|MLC-lV/sb|MYL3
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- EPR4160
- EPR4160
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- The exact immunogen used to generate this antibody is proprietary information.
- The exact immunogen used to generate this antibody is proprietary information.
- Form:
- Liquid
- Liquid
- Molecular weight:
- 21932 Da
- 21932 Da
- Purification:
- Protein A Affinity purification
- Protein A Affinity purification
- Storage buffer:
- BSA, Glycerol, Sodium chloride, Tris glycine
- BSA, Glycerol, Sodium chloride, Tris glycine
- Storage temperature:
- Store at–20 ℃.
- Store at–20 ℃.
- Shipping temperature:
- Shipped at +4 °C
- Shipped at +4 °C
- Tested applications:
- IHC-P
- IHC-P
Spécifications
Frequently Bought Together