Anti-TRIM16 Rabbit Polyclonal Antibody
Catalog # ABCA_AB72129-100UL
Supplier: Abcam
New Product
Specifications
- Antibody type:Primary
- Antigen name:EBBP
- Antigen symbol:TRIM16
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Antigen synonyms:Tripartite motif-containing protein 16|E3 ubiquitin-protein ligase TRIM16|Estrogen-responsive B box protein
- Storage buffer:BSA, Tris buffered saline
- Molecular weight:63955 Da
- Storage temperature:Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:0.2 mg/ml
- Shipping temperature:Shipped at +4 °C
- Immunogen:Synthetic Peptide within Human TRIM16. The exact immunogen used to generate this antibody is proprietary information.
- Tested applications:IP
- Purification:Immunogen Affinity purification
- Packaging:Vial
Specifications
About this item
Rabbit polyclonal to TRIM16.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: TRIM16
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Human