Anti-IPS1 Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Rabbit Polyclonal IPS1 antibody. Suitable for IP, WB and reacts with Human samples. Immunogen corresponding to Synthetic Peptide within Human PPIP5K1 aa 1350 to C-terminus.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: IPS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Taille:100 µg
- Packaging:Vial
- Western blot:Yes
- Clonality:Polyclonal
- ImmunoPrecipitation:Yes
- Purification:Affinity purification
- Antibody type:Primary
- Form:Liquid
- Isotype:IgG
- Storage temperature:Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at −20 °C long term. Avoid freeze/thaw cycle.
- Host:Rabbit
- Concentration:1 mg/ml
- Molecular weight:159521 Da
- Shipping temperature:4 °C
- Antigen name:HISPPD2A
- Cat. No.:ABCA_AB240581-100U
- Reactivity:Human
- Antigen synonyms:IP6K|KIAA0377|VIP1|PPIP5K1|Inositol hexakisphosphate and diphosphoinositol-pentakisphosphate kinase 1|Diphosphoinositol pentakisphosphate kinase 1|Histidine acid phosphatase domain-containing protein 2A|IP6 kinase|Inositol pyrophosphate synthase 1|InsP6 and PP-IP5 kinase 1|VIP1 homolog|hsVIP1
- Immunogen:Synthetic Peptide within Human PPIP5K1 aa 1350 to C-terminus. The exact immunogen used to generate this antibody is proprietary information.
- Tested applications:IP
- Antigen symbol:IPS1
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



