TRC Finasteride
TRC Finasteride
New Product
Supplier:   LGC Standards
TRC Finasteride
Supplier:  LGC Standards
New Product
Certificates
TRC Finasteride
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 77984-725
  • 77984-726
  • CAS No.:
  • 98319-26-7
  • 98319-26-7
Specifications
Product Family Options
Product Information
  • Désignation
    Taille
    Availability
    Price
  • Finasteride
    100 mg
    Log in for availability
    Log in for availability
    Each
    Each - $162.00
    $162.00
    Each
       $162.00
    Supplier Number: TRC-F342000-100MG
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    77984-725
    CAS No.
    98319-26-7
    New Product
  • Finasteride
    1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $432.00
    $432.00
    Each (1g)
       $432.00
    Supplier Number: TRC-F342000-1G
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    77984-726
    CAS No.
    98319-26-7