Chlorine Dioxide Neutralizer Solution Pack | Avantor
Chlorine Dioxide Neutralizer Solution Pack
DPD
Catalog # 77674-544
Supplier:   CHEMetrics
Fournisseur #:  A-2700
Chlorine Dioxide Neutralizer Solution Pack
Catalog # 77674-544
Supplier:  CHEMetrics
CAS Number:  
Chlorine Dioxide Neutralizer Solution Pack
Chlorine Dioxide Neutralizer Solution Pack
DPD
CAS Number:  
Price
$27.30
Log in pour la disponibilité
Pack of 6 (10ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Chromate Acidifier Solution A-2700 accessory pack supplies enough solution to perform at least 200 tests. Qty. per Pack: six 10 ml bottles.
Spécifications
Spécifications
  • Méthode chimique:
    DPD