Carbohydrazide Activator Accessory Solution Pack | Avantor
Carbohydrazide Activator Accessory Solution Pack
PDTS
Catalog # 77674-540
Supplier:   CHEMetrics
Fournisseur #:  A-1800
Carbohydrazide Activator Accessory Solution Pack
Catalog # 77674-540
Supplier:  CHEMetrics
CAS Number:  
Carbohydrazide Activator Accessory Solution Pack
Carbohydrazide Activator Accessory Solution Pack
PDTS
CAS Number:  
Price
$27.29
Log in pour la disponibilité
Pack of 6 (10ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The A-1800 Carbohydrazide Activator Accessory Solution Pack contains six 10 ml bottles of the activator solution necessary to perfrom analysis of carbohydrazide in water. This solution is found in K-1805 Carbohydrazide CHEMets®; Test Kit. This pack is meant as a refill for those who have already used their provided solution or who need to have solutions at multiple locations.
Spécifications
Spécifications
  • Méthode chimique:
    PDTS