ContactUsLinkComponent
Mast Cell IHC Control Slides
Catalogue # 77907-930
Fournisseur:  StatLab
New Product
undefined
Mast Cell IHC Control Slides
Catalogue # 77907-930
Fournisseur:  StatLab
Supplier Number:  CSM0125P
New Product
Spécifications
  • Description:
    Mast cell IHC control slides
Spécifications
À propos de cet article
Mast Cell IHC control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Stained slide is not included
    Tissue: known positive
    Special stain or antibody: Mast cell
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90 degree corners. Each pack includes specific staining protocol.