PathFlow Supa Mega Base Mold
New Product
 :   StatLab
PathFlow Supa Mega Base Mold
 :  StatLab
New Product
PathFlow Supa Mega Base Mold
PathFlow stainless steel Supa Mega base molds.
  • Reusable Stops wax leakage Stackable design for efficient storage Stainless steel for a lifetime of use
Product Family Options
  • Désignation
    Dimensions
  • PathFlow Supa Mega base mold
    36×36×10 mm
    Pack of 5
    Pack of 5 - $242.70
    $242.70
    Pack of 5
       $242.70
     : GBC-3614-05A
    New Product
    New Product
  • PathFlow Supa Mega base mold
    60×45×15 mm
    Pack of 5
    Pack of 5 - $582.21
    $582.21
    Pack of 5
       $582.21
     : GBC-6014-05A
    New Product
    New Product
  • PathFlow Supa Mega base mold
    60×45×8 mm
    Pack of 5
    Pack of 5 - $242.70
    $242.70
    Pack of 5
       $242.70
     : GBC-6014-05B