ContactUsLinkComponent
PAS (with and without Digestion) Control Slides
Catalogue # 77907-956
Fournisseur:  StatLab
New Product
undefined
PAS (with and without Digestion) Control Slides
Catalogue # 77907-956
Fournisseur:  StatLab
Supplier Number:  CS-PAS/25
New Product
Spécifications
  • Description:
    PAS (with and without digestion) control slides
Spécifications
À propos de cet article
PAS (with and without Digestion) control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Includes 1 stained slide
    Tissue: known positive
    Special stain or antibody: PAS stain
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90 degree corners. Each pack includes specific staining protocol.