Amyloid (Human) Control Slide | Avantor
Amyloid (Human) Control Slide
Amyloid (Human) control slides
Catalog # 77907-874
New Product
Supplier:   VWR International
Amyloid (Human) Control Slide
Catalog # 77907-874
Supplier:  VWR International
New Product
Amyloid (Human) Control Slide
Amyloid (Human) Control Slide
Amyloid (Human) control slides
Price
$725.23
Log in pour la disponibilité
Pack of 25
Product Details & Documents
Amyloid (Human) control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Includes one stained slide
  • Tissue: known positive special stain
  • Antibody: Congo red amyloid
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.
Spécifications
  • Désignation:
    Amyloid (Human) control slides