Amyloid (Human) Control Slide
Amyloid (Human) control slides
Catalog # 77907-874
New Product
Supplier: VWR International
Jump to:
Product Details & Documents
Amyloid (Human) control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
- Includes one stained slide
- Tissue: known positive special stain
- Antibody: Congo red amyloid
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.
Spécifications
- Désignation:Amyloid (Human) control slides