Hepatocellular Carcinoma Control Slides
Catalogue # 77907-920
Fournisseur: VWR International
New Product
Spécifications
- Description:Hepatocellular Carcinoma control slides
Spécifications
À propos de cet article
Hepatocelluar Carcinoma control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
Stained slide is not included
Tissue: known positive
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.