Fite Control Slide
Fite control slides
Catalog # 77907-904
New Product
Supplier: StatLab
: CS-FITE/25
Jump to:
Product Details & Documents
Fite control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
- Includes one stained slide
- Tissue: known positive
- Special stain or antibody: Fite's stain
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.
Spécifications
- Désignation:Fite control slides