Copper Control Slides
Copper control slides
Catalog # 77907-884
New Product
Supplier: VWR International
Jump to:
Product Details & Documents
Copper control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
Stained slide is not included
Tissue: known positive
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.
Spécifications
- Désignation:Copper control slides