ContactUsLinkComponent
Cotton Tip Applicator Stick
Cotton Tip Applicator Stick
Catalog # 77907-984
Supplier:  StatLab
New Product
Cotton Tip Applicator Stick
Catalog # 77907-984
Supplier:  StatLab
Supplier Number:  CTA-806W
New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Cotton tip applicator stick
  • Length:
    6"
Specifications
About this item
For application of tissue marking dyes and other routine uses in paraffin and frozen sectioning.
  • Splinter-free applicator stick with cotton tip
  • 6" length
  • Non sterile