Helicobacter Control Slides
Helicobacter control slides
Catalog # 77907-924
New Product
Supplier: VWR International
Jump to:
Product Details & Documents
Helicobacter control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
Includes 1 stained slide
Tissue: known positive
Special stain or antibody: Helicobacter Pylori
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90 degree corners. Each pack includes specific staining protocol.
Spécifications
- Désignation:Helicobacter control slides