AFB Acid Fast Bacteria Control Slides
77907-870
: StatLab
New Product
- Description:AFB Acid Fast Bacteria control slides
AFB Acid Fast Bacteria control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
Includes 25 unstained slides
Tissue: known positive special stain
Antibody: AFB Acid Fast Bacillus
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90 degree corners. Each pack includes specific staining protocol.