CytomeGallonovirus (CMV) IHC Control Slides
77907-890
: StatLab
New Product
- Description:CytomeGallonovirus (CMV) IHC control slides
Cytomegalovirus (CMV) IHC control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
Stained slide is not included
Tissue: known positive
Special stain or antibody: CMV
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.