Small Intestine Control Slides | Avantor
Small Intestine Control Slides
Small Intestine control slides
Catalog # 77907-968
New Product
Supplier:   VWR International
Small Intestine Control Slides
Catalog # 77907-968
Supplier:  VWR International
New Product
Small Intestine Control Slides
Small Intestine Control Slides
Small Intestine control slides
Price
$793.97
Log in pour la disponibilité
Pack of 25
Product Details & Documents
Small Intestine control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Stained slide is not included
    Tissue: Small Intestine
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.
Spécifications
  • Désignation:
    Small Intestine control slides