PathFlow Supa Mega Slim Cassettes
New Product
Supplier:   VWR International
PathFlow Supa Mega Slim Cassettes
Supplier:  VWR International
New Product
Certificates
PathFlow Supa Mega Slim Cassettes
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Supa Mega Slim cassette is being widely adopted in clinical, research, and toxicology laboratories.
  • Casting can be done in 20 minutes at −8 °C
  • Large color range available
  • Specific base molds
The Supa Mega Slim cassette offers better reagent flow and improved turnaround times, with less wax used during the embedding process.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Availability
    Price
  • PathFlow supa mega slim cassettes
    White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 100
    Box of 100 - $420.63
    $420.63
    Box of 100
       $420.63
    Spécifications:
    New Product
    New Product
  • PathFlow supa mega slim cassettes
    Green
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 100
    Box of 100 - $725.61
    $725.61
    Box of 100
       $725.61
    Spécifications:
    New Product
    New Product
  • PathFlow supa mega slim cassettes
    Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 100
    Box of 100 - $420.63
    $420.63
    Box of 100
       $420.63
    Spécifications:
    New Product
    New Product
  • PathFlow supa mega slim cassettes
    Pink
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 100
    Box of 100 - $420.63
    $420.63
    Box of 100
       $420.63
    Spécifications:
    New Product
    New Product
  • PathFlow supa mega slim cassettes
    Yellow
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 100
    Box of 100 - $420.63
    $420.63
    Box of 100
       $420.63
    Spécifications:
    New Product
    New Product
  • PathFlow supa mega slim cassettes
    Red
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 100
    Box of 100 - $420.63
    $420.63
    Box of 100
       $420.63
    Spécifications:
    New Product