Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Spécifications
- Pore size:7.0 μm
- Color:Pink
- Description:StatPrep™ LBC filters, Gyn
Spécifications
À propos de cet article
StatPrep™ ultra-thin, micro-porous polycarbonate membrane films are manufactured in a cleanroom environment.
Pink gyn filters
Compatible with standard LBC processors
A multi-step process starts with blasting membranes with accelerated heavy ions creating a highly consistent, specific pore size. Further processing ensures an ultra-thin surface that is chemically etched to assure consistent cellular results.