Pin4 (Prostatic Adenocarcinoma) IHC Control Slides
Catalog # 77907-958
Supplier: StatLab
New Product
Specifications
- Description:Pin4 (Prostatic Adenocarcinoma) IHC control slides
Specifications
About this item
One slide in each box features a serial section that has been stained using the specified staining procedure that is included in each box.
- 15 slides per pack IHC controls