ContactUsLinkComponent
CMV (Placenta) IHC Control Slide
Catalog # 77907-898
Supplier:  StatLab
New Product
undefined
CMV (Placenta) IHC Control Slide
Catalog # 77907-898
Supplier:  StatLab
Supplier Number:  CS-CMV/15
New Product
Specifications
  • Description:
    CMV (Placenta) IHC control slides
Specifications
About this item
One slide in each box features a serial section that has been stained using the specified staining procedure that is included in each box.
All boxes contain 15 slides unless noted otherwise.