PolyCut Plus Microtome Blades
New Product
Supplier:   VWR International
PolyCut Plus Microtome Blades
Supplier:  VWR International
New Product
Certificates
PolyCut Plus Microtome Blades
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
PolyCut Plus microtome blades are exclusive to StatLab and are an excellent choice for even the most difficult tissues and frozen sections.
  • Available in low profile and high profile formats
    Made in the USA
A MicroCoat® Enrichment Technology ensures the highest quality blade, one that reduces frequent blade changeout, adjustments, downtime, and waste.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Microtome blades, low profile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $331.80
    $331.80
    Pack of 50
       $331.80
    Spécifications:
    New Product
    New Product
  • Microtome blades, high profile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - $331.80
    $331.80
    Pack of 50
       $331.80
    Spécifications:
    New Product