ContactUsLinkComponent
Skin Control Slide
Catalogue # 77907-970
Fournisseur:  VWR International
New Product
undefined
Skin Control Slide
Catalogue # 77907-970
Fournisseur:  VWR International
New Product
Spécifications
  • Description:
    Skin control slides
Spécifications
À propos de cet article
Skin control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Stained slide is not included
  • Tissue: Skin
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.