Elastic (Human) Control Slides
77907-900
: StatLab
New Product
- Description:Elastic (Human) control slides
Elastic (Human) control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
Includes 1 stained slide Tissue: human known positive
Special stain or antibody: Elastic stain
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90 degree corners. Each pack includes specific staining protocol.