ClariTex Supa Mega Microscope Slides, Plain
ClariTex Supa Mega microscope slides, plain, 52×76 mm
Catalog # 77908-076
New Product
Supplier: StatLab
Fournisseur #: MAC-1400-02A
Jump to:
Product Details & Documents
Pre-cleaned and ready to use slides are an ideal time saving option.
Plain slide 1.0 to 1.2 mm square edge ground
Manufactured to ISO 8037/1
The ClariTex Supa Mega microscope slides are double the average width of a standard slide, allowing sections to be much bigger and saving pathologists time spent switching between slides and fatigue from mentally reconstructing an image from several smaller sections.
Spécifications
- Désignation:ClariTex Supa Mega microscope slides, plain
- L×l:52×76 mm