Trichrome (Human, Liver) Control Slides
Catalogue # 77907-982
Fournisseur: VWR International
New Product
Spécifications
- Description:Trichrome (Human, Liver) control slides
Spécifications
À propos de cet article
Trichrome (Human, Liver) control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
Includes one stained slide
Tissue: known positive
Special stain or antibody: Trichrome stain
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.