Spirochete Control Slides | Avantor
Spirochete Control Slides
Spirochete control slides
Catalog # 77907-974
New Product
Supplier:   VWR International
Spirochete Control Slides
Catalog # 77907-974
Supplier:  VWR International
New Product
Spirochete Control Slides
Spirochete Control Slides
Spirochete control slides
Price
$604.38
Log in pour la disponibilité
Pack of 25
Product Details & Documents
Spirochete control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Includes one stained slide
    Tissue: known positive
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.
Spécifications
  • Désignation:
    Spirochete control slides