StatPrep Adhesion CytoSlides
Supplier: VWR International
New Product
Certificates
About this item
StatPrep adhesion cytoslides are designed for use with the StatPrep liquid-based cytology collection system.
- Small and large circle formats
- Small circle: 13 mm
- Large circle: 36 mm
Frequently Bought Together