Amyloid Control Slides
77907-864
New Product
- Description:Amyloid control slides
Amyloid Control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements for amyloid.
Control slide tissue is cut at 8 microns
Intended for use with Congo Red Amyloid Special Stains
Not for use with IHC
Includes 25 unstained slides.