Large Lab Wipe | Avantor
Large Lab Wipe
Large lab wipes, 11×13"
Catalog # 77908-138
New Product
Supplier:   StatLab
Fournisseur #:  SL5790
Large Lab Wipe
Catalog # 77908-138
Supplier:  StatLab
New Product
Large Lab Wipe
Large lab wipes, 11×13"
Price
$20.28
Log in pour la disponibilité
Pack of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Use as an absorbent work surface for coverslipping or for general lab cleanup.
  • Low-linting
  • Heavy-duty, air-layered material
Spécifications
  • Désignation:
    Large lab wipes
  • L×l:
    11×13"