ContactUsLinkComponent
HistoBond®+ Supa Mega White Charged Microscope Slide
  77908-080
 :  StatLab
New Product
HistoBond®+ Supa Mega White Charged Microscope Slide
  77908-080
 :  StatLab
 :  MAD-1402-09A
New Product
  • Description:
    HistoBond®+ supa mega white charged microscope slides
  • L×W:
    52×76 mm
The autoclavable HistoBond®+ Supa Mega White Charged Slides are supplied pre-cleaned and ready for use in cardboard boxes of 50 slides.
  • Suitable for problem tissue
  • Suitable for IHC
  • White printed marking area
  • Pre-cleaned and ready to use
  • Time-saving option
  • Manufactured to ISO 8037/1
The HistoBond®+ Supa Mega White Charged Microscope Slide is made of soda-lime glass and ensures excellent specimen adhesion. The provision of a white marking area means slides can be identified with a pen, pencil, or label.