Pancreas IHC Control Sildes
Catalogue # 77907-950
Fournisseur: StatLab
New Product
Spécifications
- Description:Pancreas IHC control sildes
Spécifications
À propos de cet article
Pancreas IHC control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
Stained slide is not included
Tissue: Pancreas
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90 degree corners. Each pack includes specific staining protocol.