Alcian Blue Control Slides
77907-872
New Product
- Description:Alcian Blue control slides
Alcian Blue control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
Includes one stained slide
Tissue: known positive
Special Stain Antibody: Alcian Blue pH 1.0/2.5
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.