Order Entry
Canada
ContactUsLinkComponent
Anti-ATG7 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-ATG7 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77851-540
Supplier:  AFG Bioscience
New Product
undefined
Anti-ATG7 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77851-540
Supplier:  AFG Bioscience
Supplier Number:  A13418-50UG
New Product

Specifications

  • Antigen symbol:
    ATG7
  • Clonality:
    Recombinant Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    50 µg
  • Form:
    Liquid
  • Gene ID:
    O95352
  • Antigen synonyms:
    1810013K23Rik|Apg 7|APG7 autophagy 7 like|APG7 autophagy 7-like (S. cerevisiae)|APG7 like|APG7|S. cerevisiae|homolog of|APG7-like|APG7L|ATG 7|ATG12-activating enzyme E1 ATG7|ATG7|ATG7 autophagy related 7 homolog (S. cerevisiae)|ATG7 autophagy related 7 homolog|ATG7_HUMAN|Atg7l|Autophagy 7|Autophagy related protein 7|Autophagy-related 7 (yeast)|Autophagy-related protein 7|DKFZp434N0735|GSA 7|GSA7|hAGP7|Ubiquitin activating enzyme E1 like protein|Ubiquitin-activating enzyme E1-like protein|Ubiquitin-like modifier-activating enzyme ATG7
  • Storage buffer:
    0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
    Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Recombinant Human Ubiquitin-like modifier-activating enzyme ATG7 protein (386-676AA)
  • Purification:
    >95%, Protein G purified
  • Packaging:
    Vial

Specifications

About this item

Type:
Antigen: ATG7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human