Anti-AKR1C1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77850-974
New Product
- Antigen symbol:AKR1C1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q04828
- Antigen synonyms:2-dihydrobenzene-1|2-diol dehydrogenase|20 alpha (3 alpha) hydroxysteroid dehydrogenase|20 ALPHA HSD|20 alpha hydroxysteroid dehydrogenase|20-alpha-HSD|20-alpha-hydroxysteroid dehydrogenase|20ALPHAHSD|2ALPHAHSD|AK1C1|AK1C1_HUMAN|Aldo keto reductase family 1 member C1|aldo-keto reductase C|Aldo-keto reductase family 1 member C1|Aldo-keto reductase family|1 member 1|C9|Chlordecone reductase homolog|Chlordecone reductase homolog HAKRC|DD1|DD1/DD2|DDH|DDH1|Dihydrodiol dehydrogenase 1|Dihydrodiol dehydrogenase 1/2|20-alpha (3-alpha)-hydroxysteroid dehydrogenase|dihydrodiol dehydrogenase isoform DD1|Dihydrodiol dehydrogenase|type 1|H37|HAKRC|HBAB|Hepatic dihydrodiol dehydrogenase|High affinity hepatic bile acid-binding protein|High-affinity hepatic bile acid-binding protein|Indanol dehydrogenase|MBAB|MGC8954|Trans-1|2 dihydrobenzene 1|2 diol dehydrogenase|Type II 3 alpha hydroxysteroid dehydrogenase
- Storage buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Aldo-keto reductase family 1 member C1 protein (219-323AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: AKR1C1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human