Anti-CGGBP1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur: AFG Bioscience
New Product
Certificats
À propos de cet article
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Spécifications
- Catalog No:
- 77843-807
- 77843-806
- Antigen symbol:
- CGGBP1
- CGGBP1
- Antigen synonyms:
- 20 kDa CGG binding protein|20 kDa CGG-binding protein|CGBP1_HUMAN|CGG binding protein 1|CGG triplet repeat binding protein 1|CGG triplet repeat-binding protein 1|CGG-binding protein 1|CGGBP 1|CGGBP|Cggbp1|OTTHUMP00000213853|OTTHUMP00000213877|p20 CGG binding protein|p20 CGGBP|p20 CGGBP DNA binding protein|p20-CGGBP DNA-binding protein
- 20 kDa CGG binding protein|20 kDa CGG-binding protein|CGBP1_HUMAN|CGG binding protein 1|CGG triplet repeat binding protein 1|CGG triplet repeat-binding protein 1|CGG-binding protein 1|CGGBP 1|CGGBP|Cggbp1|OTTHUMP00000213853|OTTHUMP00000213877|p20 CGG binding protein|p20 CGGBP|p20 CGGBP DNA binding protein|p20-CGGBP DNA-binding protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UFW8
- Q9UFW8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human CGG triplet repeat-binding protein 1 protein (1-167AA)
- Recombinant Human CGG triplet repeat-binding protein 1 protein (1-167AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Spécifications
Frequently Bought Together