Anti-DUSP3 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77854-427
New Product
- Antigen symbol:DUSP3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P51452
- Antigen synonyms:Dual specificity phosphatase 3|Dual specificity protein phosphatase 3|Dual specificity protein phosphatase VHR|DUS3_HUMAN|Serine/threonine specific protein phosphatase|Vaccinia H1-related phosphatase|Vaccinia virus phosphatase VH1 related|VHR
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Dual specificity protein phosphatase 3 protein (14-96AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
0
Type:
Antigen: DUSP3
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human