Anti-CXorf1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-825
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:CXorf1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O96002
- Antigen synonyms:TMEM257|Chromosome X open reading frame 1|CXorf1|Uncharacterized protein CXorf1|CX001_HUMAN|Transmembrane Protein 257
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Putative transmembrane protein CXorf1 protein (79-111AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: CXorf1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human