Anti-DHDDS Rabbit Recombinant Polyclonal Antibody
50 µl, Vial
Catalog # 77854-144
Supplier: AFG Bioscience
Supplier #: A19397-50UL
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: DHDDS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Taille:50 µl
- Packaging:Vial
- Clonality:Recombinant Polyclonal
- Purification:Antigen affinity purified
- Form:Liquid
- Isotype:IgG
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Host:Rabbit
- Shipping temperature:Shipped on blue ice
- Cat. No.:77854-144
- Reactivity:Human
- Gene ID:Q86SQ9
- Antigen synonyms:HDSDehydrodolichyl diphosphate synthase complex subunit DHDDS|EC 2.5.1.87|Cis-isoprenyltransferase|CIT|Cis-IPTase|Cis-prenyltransferase subunit hCIT|Epididymis tissue protein Li 189m
- Immunogen:Recombinant Human Dehydrodolichyl diphosphate synthase complex subunit DHDDS protein (1-333AA)
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:DHDDS
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



