Order Entry
Canada
ContactUsLinkComponent
Anti-ASTN2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  77851-513
New Product
undefined
Anti-ASTN2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  77851-513
 :  A13359-50UG
New Product

 

  • Antigen symbol:
    ASTN2
  • Clonality:
    Recombinant Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    50 µg
  • Form:
    Liquid
  • Gene ID:
    O75129
  • Antigen synonyms:
    ASTN2_HUMAN|Astrotactin 2|Astrotactin-2|bA264C15.1|bA67K19.1|KIAA0634
  • Storage buffer:
    0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
    Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Recombinant Human Astrotactin-2 protein (108-272AA)
  • Purification:
    >95%, Protein G purified
  • Packaging:
    Vial

 

 

Type:
Antigen: ASTN2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human