Anti-BCHE Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalogue # 77851-815
Fournisseur: AFG Bioscience
New Product
Spécifications
- Antigen symbol:BCHE
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P06276
- Antigen synonyms:Cholinesterase (serum) 2|Choline esterase II|Pseudocholinesterase|Butyrylcholine esterase|E1|CHE2|Cholinesterase 1|Cholinesterase|CHE1|Acylcholine acylhydrolase|Pseudocholinesterase E1|CHLE_HUMAN|BCHE
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cholinesterase protein (311-524AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Spécifications
À propos de cet article
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: BCHE
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human