Anti-ANXA13 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
77851-170
New Product
- Antigen symbol:ANXA13
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Dog
- Size:50 µg
- Form:Liquid
- Gene ID:Q29471
- Antigen synonyms:Intestine-specific annexin|Annexin XIII|ANX13Annexin A13|ANXA13|ISA|Annexin-13
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Dog Annexin A13 protein (2-316AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: ANXA13
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog