Order Entry
Canada
ContactUsLinkComponent
Anti-FUT8 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur:  AFG Bioscience
New Product
Certificats
undefined
Anti-FUT8 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur:  AFG Bioscience
New Product

Spécifications

  • Catalog No:
  • 77842-979
  • 77842-980
  • Antigen symbol:
  • FUT8
  • FUT8
  • Antigen synonyms:
  • 6-fucosyltransferase|Alpha (1|6) fucosyltransferase|Alpha-(1|6)-fucosyltransferase|Alpha1 6FucT|alpha1-6FucT|Fucosyltransferase 8 (alpha (1|6) fucosyltransferase)|Fucosyltransferase 8|FUT8|FUT8_HUMAN|GDP fucose glycoprotein fucosyltransferase|GDP L Fuc:N acetyl beta D glucosaminide alpha1|6 fucosyltransferase|GDP-fucose--glycoprotein fucosyltransferase|GDP-L-Fuc:N-acetyl-beta-D-glucosaminide alpha1|Glycoprotein 6 alpha L fucosyltransferase|Glycoprotein 6-alpha-L-fucosyltransferase|MGC26465
  • 6-fucosyltransferase|Alpha (1|6) fucosyltransferase|Alpha-(1|6)-fucosyltransferase|Alpha1 6FucT|alpha1-6FucT|Fucosyltransferase 8 (alpha (1|6) fucosyltransferase)|Fucosyltransferase 8|FUT8|FUT8_HUMAN|GDP fucose glycoprotein fucosyltransferase|GDP L Fuc:N acetyl beta D glucosaminide alpha1|6 fucosyltransferase|GDP-fucose--glycoprotein fucosyltransferase|GDP-L-Fuc:N-acetyl-beta-D-glucosaminide alpha1|Glycoprotein 6 alpha L fucosyltransferase|Glycoprotein 6-alpha-L-fucosyltransferase|MGC26465
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9BYC5
  • Q9BYC5
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Alpha-(1,6)-fucosyltransferase protein (247-377AA)
  • Recombinant Human Alpha-(1,6)-fucosyltransferase protein (247-377AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage temperature:
  • Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
  • Shipping temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Spécifications

Frequently Bought Together