Anti-CCL8 Rabbit Recombinant Polyclonal Antibody
77852-495
New Product
- Antigen symbol:CCL8
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P80075
- Antigen synonyms:CCL8_HUMAN|HC14|MCP-2|MCP-2(6-76)|Monocyte chemoattractant protein 2|Monocyte chemotactic protein 2|SCYA10|SCYA8|Small-inducible cytokine A8
- Storage buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human C-C motif chemokine 8 protein (23-99AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: CCL8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human