Anti-ASNS Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalogue # 77851-493
Fournisseur: AFG Bioscience
New Product
Spécifications
- Antigen symbol:ASNS
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P08243
- Antigen synonyms:ASNS_HUMAN|ASNSD|Asparagine synthetase [glutamine-hydrolyzing]|Asparagine synthetase|Cell cycle control protein TS11|Glutamine dependent asparagine synthetase 3|Glutamine dependent asparagine synthetase|Glutamine hydrolyzing|Glutamine-dependent asparagine synthetase|OTTHUMP00000024510|OTTHUMP00000204938|OTTHUMP00000204939|OTTHUMP00000204940|OTTHUMP00000204941|OTTHUMP00000204942|TS11|TS11 cell cycle control protein
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Asparagine synthetase [glutamine-hydrolyzing] protein (364-551AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Spécifications
À propos de cet article
Type:
Antigen: ASNS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human