Anti-KCNS2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000 to 1:5000, IF:1:50 to 1:200
Type:
Antigen: KCNS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77845-150
- 77845-149
- Antigen symbol:
- KCNS2
- KCNS2
- Antigen synonyms:
- Delayed rectifier K(+) channel alpha subunit 2|Delayed-rectifier K(+) channel alpha subunit 2|Kcns2|KCNS2_HUMAN|KIAA1144|KV9.2|Potassium voltage gated channel subfamily S member 2|Potassium voltage-gated channel subfamily S member 2|Voltage gated potassium channel subunit Kv9.2|Voltage-gated potassium channel subunit Kv9.2
- Delayed rectifier K(+) channel alpha subunit 2|Delayed-rectifier K(+) channel alpha subunit 2|Kcns2|KCNS2_HUMAN|KIAA1144|KV9.2|Potassium voltage gated channel subfamily S member 2|Potassium voltage-gated channel subfamily S member 2|Voltage gated potassium channel subunit Kv9.2|Voltage-gated potassium channel subunit Kv9.2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9ULS6
- Q9ULS6
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Potassium voltage-gated channel subfamily S member 2 protein (1-183AA)
- Recombinant Human Potassium voltage-gated channel subfamily S member 2 protein (1-183AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together