Anti-CEP290 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
77852-989
New Product
- Antigen symbol:CEP290
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O15078
- Antigen synonyms:3H11AG|Bardet-Biedl syndrome 14 protein|BBS14|Cancer/testis antigen 87|CE290_HUMAN|Centrosomal protein 290|Centrosomal protein 290kDa|Centrosomal protein of 290 kDa|Cep290|CT87|CTCL tumor antigen se2 2|FLJ13615|FLJ21979|JBTS5|JBTS6|KIAA0373|LCA10|Meckel syndrome|type 4|MKS4|Monoclonal 3H11 antigen|Nephrocystin 6|Nephrocystin-6|NPHP6|POC3|POC3 centriolar protein homolog|Prostate cancer antigen T21|rd16|SLSN6 1|2|5|SLSN6|Tumor antigen se2-2
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human Centrosomal protein of 290 kDa protein (86-156AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: CEP290
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human