Anti-BAIAP3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77851-780
Supplier: AFG Bioscience
New Product
Specifications
- Antigen symbol:BAIAP3
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O94812
- Antigen synonyms:BAI1 associated protein 3|BAI1-associated protein 3|Baiap3|BAIP3_HUMAN|BAP2|BAP3
- Storage buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Recombinant Human BAI1-associated protein 3 protein (131-257AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: BAIAP3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human