Anti-MAPT (phospho-Ser404) Rabbit Polyclonal Antibody
Catalogue # 77848-938
Fournisseur: AFG Bioscience
New Product
Spécifications
- Antigen symbol:MAPT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P10636
- Antigen synonyms:AI413597|AW045860|DDPAC|FLJ31424|FTDP 17|G protein beta1/gamma2 subunit interacting factor 1|MAPTL|MGC134287|MGC138549|MGC156663|Microtubule associated protein tau|Microtubule associated protein tau isoform 4|Microtubule-associated protein tau|MSTD|Mtapt|MTBT1|MTBT2|Neurofibrillary tangle protein|Paired helical filament tau|Paired helical filament-tau|PHF tau|PHF-tau|PPND|PPP1R103|Protein phosphatase 1, regulatory subunit 103|pTau|RNPTAU|TAU|TAU_HUMAN|Tauopathy and respiratory failure|Tauopathy and respiratory failure, included
- Modification:phospho-Ser404
- Storage buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping temperature:Shipped on blue ice
- Immunogen:Peptide sequence around phosphorylation site of serine 404 (D-T-S(p)-P-R) derived from Human Tau.
- Purification:Antigen affinity purified
- Packaging:Vial
Spécifications
À propos de cet article
Recommended dilution: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, IF: 1:100 to 1:200
Type:
Antigen: MAPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat